missing translation for 'onlineSavingsMsg'
Learn More

AIP/ARA9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£292.00 - £488.00

Specifications

Antigen AIP/ARA9
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18260925
View Documents
Novus Biologicals
NBP2-55628
100 μL
£488.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18667128
View Documents
Novus Biologicals
NBP2-55628-25ul
25 μL
£292.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

AIP/ARA9 Polyclonal specifically detects AIP/ARA9 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

AIP/ARA9
Polyclonal
Rabbit
Apoptosis, Hypoxia, Neuroscience, Neurotransmission
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
9049
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MTDEEKAKAVPLIHQEGNRLYREGHVKEAAAKYYDAIACLKNLQMKEQPGSPEWIQLDKQITPLLLNYCQCKLVVEEYYEVLDHCSSILNKYDDNVKAYFKRGKAHAAVWNAQEAQADFAKVLELDPALAPVVSRELQALE
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Immunocytochemistry, Immunofluorescence
Unconjugated
RUO
Human
ARA9, FKBP16, FKBP37, SMTPHN, XAP2, XAP-2
AIP
IgG
Affinity Purified
Videoer
SDS
Dokumenter

Dokumenter

Certifikater

For Research Use Only

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.