Learn More
Aiolos Monoclonal Antibody (8B2), eFluor™ 660, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£111.00 - £298.00
Specifications
Antigen | Aiolos |
---|---|
Clone | 8B2 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | eFluor 660 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15331930
|
Affymetrix eBioscience
50-5789-82 |
100 μg |
£298.00
100µg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15321930
|
Affymetrix eBioscience
50-5789-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This gene encodes a member of the Ikaros family of zinc-finger proteins. Three members of this protein family (Ikaros, Aiolos and Helios) are hematopoietic-specific transcription factors involved in the regulation of lymphocyte development. This gene product is a transcription factor that is important in the regulation of B lymphocyte proliferation and differentiation. Both Ikaros and Aiolos can participate in chromatin remodeling. Regulation of gene expression in B lymphocytes by Aiolos is complex as it appears to require the sequential formation of Ikaros homodimers, Ikaros/Aiolos heterodimers, and Aiolos homodimers. At least six alternative transcripts encoding different isoforms have been described.
This gene encodes a member of the Ikaros family of zinc-finger proteins. Three members of this protein family (Ikaros, Aiolos and Helios) are hematopoietic-specific transcription factors involved in the regulation of lymphocyte development. This gene product is a transcription factor that is important in the regulation of B lymphocyte proliferation and differentiation. Both Ikaros and Aiolos can participate in chromatin remodeling. Regulation of gene expression in B lymphocytes by Aiolos is complex as it appears to require the sequential formation of Ikaros homodimers, Ikaros/Aiolos heterodimers, and Aiolos homodimers. At least six alternative transcripts encoding different isoforms have been described.Specifications
Aiolos | |
Flow Cytometry | |
eFluor 660 | |
Mouse | |
Mouse | |
22780 | |
IgG1 κ | |
Affinity chromatography | |
Ikzf3 |
8B2 | |
Monoclonal | |
Liquid | |
RUO | |
O08900 | |
Ikzf3 | |
Primary | |
Antibody |
For Research Use Only.