Learn More
Aiolos Monoclonal Antibody (8B2), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£179.00 - £403.00
Specifications
Antigen | Aiolos |
---|---|
Clone | 8B2 |
Concentration | 0.5 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15287467
|
Affymetrix eBioscience
14-5789-80 |
25 μg |
£179.00
25µg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15297467
|
Affymetrix eBioscience
14-5789-82 |
100 μg |
£403.00
100µg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: This 8B2 monoclonal antibody reacts with mouse Aiolos, a 58-kDa member of the Ikaros family of zinc finger transcription factors. Expression of Aiolos is restricted to the lymphoid lineage, with B cells exhibiting the highest levels of expression, followed by NK and T cells. Studies demonstrate that Aiolos forms homodimers or heterodimerizes with Ikaros proteins. Moreover, Aiolos exhibits tyrosine phosphorylation and nuclear localization in response to IL-2 stimulation in T cells. Aiolos plays a role in IL-2-mediated anti-apoptosis by controlling Bcl-2 expression. Finally, Aiolos is involved in B cell development, with upregulation of this transcription factor observed at the pre-BI to pre-BII transition in response to pre-B cell receptor signaling. Applications Reported: This 8B2 antibody has been reported for use in immunoblotting/western blotting. Applications Tested: This 8B2 antibody has been tested by immunoblotting of reduced cell lysate prepared from mouse splenocytes. This can be used at less than or equal to 5 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 µm post-manufacturing filtered.
This gene encodes a member of the Ikaros family of zinc-finger proteins. Three members of this protein family (Ikaros, Aiolos and Helios) are hematopoietic-specific transcription factors involved in the regulation of lymphocyte development. This gene product is a transcription factor that is important in the regulation of B lymphocyte proliferation and differentiation. Both Ikaros and Aiolos can participate in chromatin remodeling. Regulation of gene expression in B lymphocytes by Aiolos is complex as it appears to require the sequential formation of Ikaros homodimers, Ikaros/Aiolos heterodimers, and Aiolos homodimers. At least six alternative transcripts encoding different isoforms have been described.Specifications
Aiolos | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
5830411O07Rik; AIO; AIOLOS; IKAROS family zinc finger 3; IKAROS family zinc finger 3 (Aiolos); ikaros family zinc finger protein 3; Ikzf3; Zfpn1a3; zinc finger DNA binding protein Aiolos; zinc finger protein Aiolos; zinc finger protein, subfamily 1A, 3 (Aiolos); Znfn1a3 | |
Ikzf3 | |
Primary | |
4° C | |
Ikzf3 |
8B2 | |
Western Blot | |
Unconjugated | |
Mouse | |
Mouse | |
O08900 | |
22780 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.