Learn More
Invitrogen™ Aiolos Monoclonal Antibody (8B2), eFluor™ 660, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
£331.00
Specifications
| Antigen | Aiolos |
|---|---|
| Clone | 8B2 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
This 8B2 MAb reacts with mouse Aiolos, a 58-kDa member of the Ikaros family of zinc finger transcription factors. Expression of Aiolos is restricted to the lymphoid lineage, with B cells exhibiting the highest levels of expression, followed by NK and T cells. Studies demonstrate that Aiolos forms homodimers or heterodimerizes with Ikaros proteins. Moreover, Aiolos exhibits tyrosine phosphorylation and nuclear localization in response to IL-2 stimulation in T cells. Aiolos plays a role in IL-2-mediated anti-apoptosis by controlling Bcl-2 expression. Finally, Aiolos is involved in B cell development, with upregulation of this transcription factor observed at the pre-BI to pre-BII transition in response to pre-B cell receptor signaling.
This gene encodes a member of the Ikaros family of zinc-finger proteins. Three members of this protein family (Ikaros, Aiolos and Helios) are hematopoietic-specific transcription factors involved in the regulation of lymphocyte development. This gene product is a transcription factor that is important in the regulation of B lymphocyte proliferation and differentiation. Both Ikaros and Aiolos can participate in chromatin remodeling. Regulation of gene expression in B lymphocytes by Aiolos is complex as it appears to require the sequential formation of Ikaros homodimers, Ikaros/Aiolos heterodimers, and Aiolos homodimers. At least six alternative transcripts encoding different isoforms have been described.Specifications
| Aiolos | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| O08900 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| 8B2 | |
| Flow Cytometry | |
| eFluor 660 | |
| Mouse | |
| Mouse | |
| 22780 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Ikzf3 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.