missing translation for 'onlineSavingsMsg'
Learn More

Aiolos/IKZF3 Antibody, PerCP, Novus Biologicals™

Product Code. 18421512
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18421512 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18421512 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP224495PCP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Aiolos/IKZF3 Polyclonal antibody specifically detects Aiolos/IKZF3 in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aiolos/IKZF3
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias AIO, AIOLOS, IKAROS family zinc finger 3 (Aiolos), Ikaros family zinc finger protein 3, zinc finger protein Aiolos, ZNFN1A3subfamily 1A, 3 (Aiolos)
Host Species Rabbit
Immunogen A portion of amino acids 350-400 of human IKZF3 was used as the immunogen.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Research
Primary or Secondary Primary
Gene ID (Entrez) 22806
Target Species Human, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.