missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aiolos/IKZF3 Antibody (OTI7E11), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71698H
This item is not returnable.
View return policy
Description
Aiolos/IKZF3 Monoclonal specifically detects Aiolos/IKZF3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Aiolos/IKZF3 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| IKZF3 | |
| Recombinant protein fragment corresponding to amino acids 1-301 of human Aiolos/IKZF3 (NP_036613) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI7E11 | |
| Western Blot, Immunohistochemistry | |
| AIO, AIOLOS, IKAROS family zinc finger 3 (Aiolos), Ikaros family zinc finger protein 3, zinc finger protein Aiolos, ZNFN1A3subfamily 1A, 3 (Aiolos) | |
| Mouse | |
| Affinity Purified | |
| Chromatin Research | |
| 22806.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction