missing translation for 'onlineSavingsMsg'
Learn More

AIMS™ ATS-3 General Rodent Tattoo System Pigment

Product Code. p-4278522
Encompass_Preferred
Click to view available options
Color:
Black
Capacity (English):
2 oz.
Capacity (Metric):
59.14 mL
This item is not returnable. View return policy

Product Code. 15314911

Brand: AIMS™ 14370167

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Will not alter the physiology of animals or contaminate other lab animals in the facility

  • Safe
  • Nontoxic

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance.

Specifications

Capacity (English) 2 oz.
Capacity (Metric) 59.14 mL
Color Black
For Use With (Application) In-vivo and in-vitro Toxicology Assay
Type Tattoo Pigment
For Use With (Equipment) ATS-3 general rodent tattoo system
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.