missing translation for 'onlineSavingsMsg'
Learn More

AIMS™ ATS-3 General Rodent Tattoo System Pigment

Product Code. 15314911
Encompass_Preferred
Change view
Click to view available options
Color:
Black
Capacity (English):
2 oz.
Capacity (Metric):
59.14 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Capacity (English) Capacity (Metric)
15314911 Black 2 oz. 59.14 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15314911 Supplier AIMS™ Supplier No. 14370167
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Will not alter the physiology of animals or contaminate other lab animals in the facility

  • Safe
  • Nontoxic

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance.

Specifications

Capacity (English) 2 oz.
Capacity (Metric) 59.14 mL
Color Black
For Use With (Application) In-vivo and in-vitro Toxicology Assay
Type Tattoo Pigment
For Use With (Equipment) ATS-3 general rodent tattoo system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.