missing translation for 'onlineSavingsMsg'
Learn More

AIMP2 Antibody, DyLight 650, Novus Biologicals™

Product Code. 18419956 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18419956 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18419956 Supplier Novus Biologicals Supplier No. NBP281885C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AIMP2 Polyclonal antibody specifically detects AIMP2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen AIMP2
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias aminoacyl tRNA synthase complex-interacting multifunctional protein 2, aminoacyl tRNA synthetase complex-interacting multifunctional protein 2, ARS-interacting multi-functional protein 2, JTV1JTV-1, Multisynthase complex auxiliary component p38, multisynthetase complex auxiliary component p38, p38, PRO0992, Protein JTV-1
Host Species Rabbit
Immunogen AIMP2 antibody was raised against a 16 amino acid peptide near the center of human AIMP2. The immunogen is located within amino acids 150 - 200 of AIMP2. Amino Acid Squence: FGEQNKKQPRQDYQL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 7965
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.