missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIM2 Antibody (10M5G5), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-27354R
This item is not returnable.
View return policy
Description
AIM2 Monoclonal specifically detects AIM2 in Human samples. It is validated for Western Blot.
Specifications
| AIM2 | |
| Monoclonal | |
| DyLight 550 | |
| O14862 | |
| AIM2 | |
| This antibody was raised against AIM2 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 10M5G5 | |
| Western Blot | |
| absent in melanoma 2PYHIN4, interferon-inducible protein AIM2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 9447 | |
| Store at 4C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction