missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIM2 Antibody (10M5G5) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80565
This item is not returnable.
View return policy
Description
AIM2 Monoclonal specifically detects AIM2 in Human samples. It is validated for Western Blot.
Specifications
| AIM2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| Western Blot 2-5ug/ml | |
| absent in melanoma 2PYHIN4, interferon-inducible protein AIM2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 9447 | |
| Human | |
| Purified |
| Western Blot | |
| 10M5G5 | |
| Unconjugated | |
| PBS | |
| AIM2 | |
| This antibody was raised against AIM2 | |
| 0.1 mg | |
| Primary | |
| Human | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction