missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIM2 Antibody (10M2B3), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-27355AF532
This item is not returnable.
View return policy
Description
AIM2 Monoclonal specifically detects AIM2 in Human samples. It is validated for Western Blot.
Specifications
| AIM2 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| AIM2 | |
| This antibody was raised against recombinant human AIM2 protein | |
| 0.1 mL | |
| 9447.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| 10M2B3 | |
| Western Blot | |
| absent in melanoma 2PYHIN4, interferon-inducible protein AIM2 | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Human | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction