Learn More
Invitrogen™ AIM Recombinant Rabbit Monoclonal Antibody (11H48L16)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ 703558
Description
This antibody is predicted to react with Monkey, Rat. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
CD5L, also known as CD5 antigen-like, is a protein that is expressed on the surface of various immune cells, including T cells, B cells, and natural killer cells. It is involved in the regulation of immune responses and is being studied as a potential therapeutic target in various diseases, including autoimmune disorders, cancer, and infectious diseases. CD5L is known to modulate T cell activation and cytokine production, and has been shown to have anti-inflammatory effects.
Specifications
| AIM | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9QWK4 | |
| Cd5l | |
| Peptides corresponding to Mouse AIM [1) aa42-aa54 2) aa271-aa283]. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| Immunocytochemistry | |
| 11H48L16 | |
| Unconjugated | |
| Cd5l | |
| 1/6; AAC-11; AI047839; AIM; AIM/Spalpha; API6; apoptosis inhibitor 6; apoptosis inhibitor expressed by macrophages; apoptosis inhibitor of macrophage; APOPTOSIS INHIBITOR OF MACROPHAGES; Apoptosis inhibitory 6; CD5 antigen like (scavenger receptor cysteine rich family); CD5 antigen-like; CD5 antigen-like (scavenger receptor cysteine rich family); CD5 molecule like; Cd5 molecule-like; CD5L; CT2; CT-2; hAIM; Highly similar to ANTIGEN WC1.1 [Bos taurus]; igM-associated peptide; mAIM; Pdp; Pdp 1/6; PRO229; SCAVENGER RECEPTOR CYSTEINE-RICH FAMILY; Spalpha; Sp-alpha; UNQ203/PRO229 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 11801 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.