missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AIM Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584779
This item is not returnable.
View return policy
Description
Immunogen sequence: LGCGAASGTP SGILYEPPAE KEQKVLIQSV SCTGTEDTLA QCEQEEVYDC Highest antigen sequence indentity to the following orthologs: Mouse - 63%, Rat - 59%.
CD5L, also known as CD5 antigen-like, is a protein that is expressed on the surface of various immune cells, including T cells, B cells, and natural killer cells. It is involved in the regulation of immune responses and is being studied as a potential therapeutic target in various diseases, including autoimmune disorders, cancer, and infectious diseases. CD5L is known to modulate T cell activation and cytokine production, and has been shown to have anti-inflammatory effects.
Specifications
| AIM | |
| Polyclonal | |
| Unconjugated | |
| Cd5l | |
| 1/6; AAC-11; AI047839; AIM; AIM/Spalpha; API6; apoptosis inhibitor 6; apoptosis inhibitor expressed by macrophages; apoptosis inhibitor of macrophage; APOPTOSIS INHIBITOR OF MACROPHAGES; Apoptosis inhibitory 6; CD5 antigen like (scavenger receptor cysteine rich family); CD5 antigen-like; CD5 antigen-like (scavenger receptor cysteine rich family); CD5 molecule like; Cd5 molecule-like; CD5L; CT2; CT-2; hAIM; Highly similar to ANTIGEN WC1.1 [Bos taurus]; igM-associated peptide; mAIM; Pdp; Pdp 1/6; PRO229; SCAVENGER RECEPTOR CYSTEINE-RICH FAMILY; Spalpha; Sp-alpha; UNQ203/PRO229 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 922 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43866 | |
| Cd5l | |
| Recombinant protein corresponding to Human CD5L. Recombinant protein control fragment (Product #RP-103668). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction