missing translation for 'onlineSavingsMsg'
Learn More

Gypenoside XVII, MedChemExpress

Product Code. 16419018
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16419018

Brand: Medchem Express HYN0553

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Gypenoside XVII, a novel phytoestrogen belonging to the gypenosides, can activate estrogen receptors.

Gypenoside XVII, a novel phytoestrogen belonging to the gypenosides, can activate estrogen receptors.

Specifications

Chemical Name or Material Gypenoside XVII
CAS 80321-69-3
Color White
Purity Grade Notes Research
Molecular Formula C48H82O18
Quantity 10 mg
Synonym Gynosaponin S
Solubility Information DMSO : ≥ 100 mg/mL (105.58 mM) ∣H2O : 41.67 mg/mL (44.00 mM; Need ultrasonic)
SMILES C[C@]([C@@](C[C@H]1O)([H])[C@]2(CC[C@@H]3O[C@]([C@@H]([C@@H](O)[C@@H]4O)O)([H])O[C@@H]4CO)C)(CC[C@@]2([H])C3(C)C)[C@]5([C@@]1([H])[C@]([C@@](CC/C=C(C)/C)(C)O[C@@H]([C@@H]([C@@H](O)[C@@H]6O)O)O[C@@H]6CO[C@@H]([C@@H]([C@@H](O)[C@@H]7O)O)O[C@@H]7CO)([H])CC5)C
Molecular Weight (g/mol) 947.15
Formula Weight 947.15
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.