missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14468-1-AP-150UL
This item is not returnable.
View return policy
Description
Androgen-induced 1 (AIG1) is a novel Pirh2-interacting protein, which was initially cloned from human dermal papilla cells and is evolutionally conserved from drosophila through to humans. It is a membrane protein and belongs to the protein family containing FAR-17a domain. AIG1 may play a role in androgen-regulated growth of hair follicles. AIG1 could serve as a new biomarker for the diagnosis and prognostic evaluation of HCCs and may be a novel candidate tumor suppressor.Specifications
| AIG1 | |
| Polyclonal | |
| Unconjugated | |
| AIG1 | |
| AIG 1, AIG1, androgen induced 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 292486, 51390, 66253 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9D8B1, Q9NVV5 | |
| AIG1 | |
| AIG1 Fusion Protein Ag5836 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction