missing translation for 'onlineSavingsMsg'
Learn More

AIFM3 Antibody, DyLight 405, Novus Biologicals™

Product Code. 18403533 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18403533 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18403533 Supplier Novus Biologicals Supplier No. NBP176889V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AIFM3 Polyclonal antibody specifically detects AIFM3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen AIFM3
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias AIFLFLJ30473, apoptosis-inducing factor 3, apoptosis-inducing factor like, apoptosis-inducing factor, mitochondrion-associated, 3, Apoptosis-inducing factor-like protein, FLJ45137
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide from near the amino terminus of human AIFM3. The immunogen is located within the first 50 amino acids of AIFM3. Amino Acid Squence: GKGSPRAYQGNGTARH
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 150209
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.