missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIFM3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92837-0.02ml
This item is not returnable.
View return policy
Description
AIFM3 Polyclonal antibody specifically detects AIFM3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| AIFM3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| AIFLFLJ30473, apoptosis-inducing factor 3, apoptosis-inducing factor like, apoptosis-inducing factor, mitochondrion-associated, 3, Apoptosis-inducing factor-like protein, FLJ45137 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human AIFM3 (NP_001018070.1). MGGCFSKPKPVELKIEVVLPEKERGKEELSASGKGSPRAYQGNGTARHFHTEERLSTPHPYPSPQDCVEAAVCHVKDLENGQMREVELGWGKVLLVKDNGEFHALGHKCPHYGAPLVKGVLSRGRVRCPWHGACFNISTGDLEDFPGLDSLHKFQVKIEKEKVYVRASKQALQLQRRTKVMAKCISPSAGYSSSTNVLIV | |
| 0.02 mL | |
| Apoptosis, Cell Biology | |
| 150209 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction