missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIF Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA548108
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
AIF Polyclonal antibody specifically detects AIF in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.Specifications
AIF | |
Polyclonal | |
Unconjugated | |
AIFM1 | |
AG1; AGR1; Aif; AIFM1; AIFsh2; apoptosis inducing factor mitochondria associated 1; apoptosis inducing factor, mitochondria associated 1; apoptosis-inducing factor 1, mitochondrial; apoptosis-inducing factor, mitochondrion-associated 1; apoptosis-inducing factor, mitochondrion-associated, 1; CMT2D; CMTX4; COWCK; COXPD6; DFNX5; ER protein 18; ER protein 19; ERP18; ERP19; hAG-1; harlequin; Hq; hTLP19; MGC111425; mitochondrial apoptosis-inducing factor 1; NADMR; NAMSD; pcd8; PDCD8; PDIA16; programmed cell death 8 (apoptosis-inducing factor); programmed cell death protein 8; RP3-438D16.2; striatal apoptosis-inducing factor; testicular secretory protein Li 4; TLP19; UNQ713/PRO1376 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26926, 83533, 9131 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O95831, Q9JM53, Q9Z0X1 | |
AIFM1 | |
E. coli-derived recombinant human AIF Glu121-Asp613 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |