missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIF, p Ser116 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-23346
This item is not returnable.
View return policy
Description
AIF Polyclonal antibody specifically detects AIF in Human samples. It is validated for Western Blot
Specifications
| AIF | |
| Polyclonal | |
| Western Blot 1:1000 | |
| AIFapoptosis-inducing factor 1, mitochondrial, apoptosis-inducing factor, mitochondrion-associated, 1, COXPD6, MGC111425, PDCD8striatal apoptosis-inducing factor, programmed cell death 8 (apoptosis-inducing factor), Programmed cell death protein 8 | |
| Phospho-AIF (Ser-116) synthetic peptide (coupled to carrier) corresponding to amino acids surrounding Ser-116 in human AIF. This site is conserved in rat and mouse AIF, but the amino acids surrounding the site are not well conserved. The site is not found in AIFM2 and AIFM3. [UniProt# O95831] | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, Inhibitors Activators and Regulators, Mitochondrial Markers, Tumor Suppressors | |
| 9131 | |
| Store at -20°C. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS, 1 mg/ml BSA and 50% glycerol | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction