missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIF Antibody (22E9), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59432G
This item is not returnable.
View return policy
Description
AIF Monoclonal antibody specifically detects AIF in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| AIF | |
| Monoclonal | |
| DyLight 488 | |
| AIFapoptosis-inducing factor 1, mitochondrial, apoptosis-inducing factor, mitochondrion-associated, 1, COXPD6, MGC111425, PDCD8striatal apoptosis-inducing factor, programmed cell death 8 (apoptosis-inducing factor), Programmed cell death protein 8 | |
| Recombinant human AIF (98-609aa) purified from E. coli | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Chromatin Research, Inhibitors Activators and Regulators, Mitochondrial Markers, Tumor Suppressors | |
| 9131 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, ELISA, Immunofluorescence | |
| 22000000000 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction