missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIF-1/Iba1 Rabbit anti-Human, Clone: 31, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90271-50UL
This item is not returnable.
View return policy
Description
AIF-1/Iba1 Monoclonal specifically detects AIF-1/Iba1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| AIF-1/Iba1 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 31 | |
| ELISA 1:5000-1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunohistochemistry-Paraffin 1:100-1:500 | |
| AIF-1IRT1, allograft inflammatory factor 1, G1, IBA1Em:AF129756.17, IbaI, interferon gamma responsive transcript, Ionized calcium-binding adapter molecule 1, IRT-1, Protein G1 | |
| Recombinant Human AIF-1/Iba1 Protein | |
| 50 μg | |
| Autoimmune Diseases, Cancer, Cell Biology, Cell Cycle and Replication, Immunology, Inflammation, Lipid and Metabolism, Microglia Markers, Neuroscience | |
| 199 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction