missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIF-1/Iba1 Antibody (rAIF1/1909), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-75760AF594
This item is not returnable.
View return policy
Description
AIF-1/Iba1 Monoclonal specifically detects AIF-1/Iba1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| AIF-1/Iba1 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| AIF1 | |
| Purified recombinant human AIF-1/Iba1 protein (around aa 1-146) (exact sequence is proprietary) (Uniprot: P55008) | |
| 0.1 mL | |
| Autoimmune Diseases, Cancer, Cell Biology, Cell Cycle and Replication, Immunology, Inflammation, Lipid and Metabolism, Microglia Markers, Neuroscience | |
| AIF1 is a cytoplasmic, calcium-binding protein that is thought to play a role in macrophage activation and function. AIF1, containing two EF domains, is induced by cytokines and Interferons. In an unstimulated state, AIF1 colocalizes with actin, and upon stimulation, translocates to lamellipodia. It is also a marker of human microglia and is expressed by macrophages in injured skeletal muscle. The gene encoding AIF1 resides in the tumor necrosis factor (TNF) cluster of genes, located in the region represented by the human major histocompatibility complex (MHC). | |
| Store at 4°C in the dark. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| rAIF1/1909 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| AIF-1, allograft inflammatory factor 1, G1, IBA1, IbaI, interferon gamma responsive transcript, Ionized calcium-binding adapter molecule 1, IRT1, IRT-1, Protein G1 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction