missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIF-1/Iba1 Antibody (CL10936) [DyLight 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21185DL594
This item is not returnable.
View return policy
Description
AIF-1/Iba1 Monoclonal antibody specifically detects AIF-1/Iba1 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| AIF-1/Iba1 | |
| Monoclonal | |
| DyLight 594 | |
| AIF-1, allograft inflammatory factor 1, G1, IBA1, IbaI, interferon gamma responsive transcript, Ionized calcium-binding adapter molecule 1, IRT1, IRT-1, Protein G1 | |
| This antibody was developed using a synthetic peptide derived from P55008, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Autoimmune Diseases, Cancer, Cell Biology, Cell Cycle and Replication, Immunology, Inflammation, Lipid and Metabolism, Microglia Markers, Neuroscience | |
| 199 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL10936 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction