Learn More
AIF-1/Iba1 Antibody (CL10930) [DyLight 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-21184UV
Product Code. 30293933
  • £558.00 / 0.10mL
Description
AIF-1/Iba1 Monoclonal antibody specifically detects AIF-1/Iba1 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
AIF-1/Iba1
Monoclonal
DyLight 350
AIF-1, allograft inflammatory factor 1, G1, IBA1, IbaI, interferon gamma responsive transcript, Ionized calcium-binding adapter molecule 1, IRT1, IRT-1, Protein G1
This antibody was developed using a synthetic peptide derived from P55008, with the exact immunogen sequence remaining proprietary.
0.1 mL
Autoimmune Diseases, Cancer, Cell Biology, Cell Cycle and Replication, Immunology, Inflammation, Lipid and Metabolism, Microglia Markers, Neuroscience
199
Store at 4⪚C in the dark.
IgG2b
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL10930
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified