missing translation for 'onlineSavingsMsg'
Learn More

AIF-1/Iba1 Antibody (031), Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18343240 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18343240 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18343240 Supplier Novus Biologicals Supplier No. NBP290271AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

AIF-1/Iba1 Monoclonal specifically detects AIF-1/Iba1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AIF-1/Iba1
Applications ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 31
Conjugate Alexa Fluor 350
Formulation 50 mM Sodium Borate
Gene Alias AIF-1, allograft inflammatory factor 1, G1, IBA1, IbaI, interferon gamma responsive transcript, Ionized calcium-binding adapter molecule 1, IRT1, IRT-1, Protein G1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human AIF-1/Iba1 (P55008-1; Met1-Pro147).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autoimmune Diseases, Cancer, Cell Biology, Cell Cycle and Replication, Immunology, Inflammation, Lipid and Metabolism, Microglia Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 199
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.