missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIDA Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23724-1-AP-20UL
This item is not returnable.
View return policy
Description
AIDA acts as a ventralizing factor during embryogenesis. Inhibits axin-mediated JNK activation by binding axin and disrupting axin homodimerization. This in turn antagonizes a Wnt/beta-catenin-independent dorsalization pathway activated by AXIN/JNK-signaling.Specifications
| AIDA | |
| Polyclonal | |
| Unconjugated | |
| AIDA | |
| AIDA, C1orf80, RP11 378J18.7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 108909, 64853 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8C4Q6, Q96BJ3 | |
| AIDA | |
| AIDA Fusion Protein Ag20588 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction