missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AID Polyclonal Antibody

AID Polyclonal Antibody
Brand: Invitrogen PA586825
This item is not returnable.
View return policy
Description
AID Polyclonal Antibody for Western Blot
Specifications
| AID | |
| Polyclonal | |
| Unconjugated | |
| Aicda | |
| activation induced deaminase; activation-induced cytidine deaminase; Aicda; AID; Arp2; CDA2; Cytidine aminohydrolase; epididymis secretory protein Li 284; HEL-S-284; HIGM2; integrated into Burkitt's lymphoma cell line Ramos; single-stranded DNA cytosine deaminase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 399679, 57379 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9GZX7 | |
| Aicda | |
| A synthetic peptide corresponding to residues in Human AICDA. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction