missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AID Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588506
This item is not returnable.
View return policy
Description
Immunogen sequence: NVRWAKGRRE TYLCYVVKRR DSATSFSLDF GYLRNKNGCH VELLFLRYIS DWDLDPGRCY RVTWFTSWSP CYDCARHVAD FLRGNPNLSL RIFTARLYFC EDRKAEPEGL RRLH; Positive Samples: Mouse brain; Cellular Location: Cytoplasm, Nucleus.
This gene encodes a RNA-editing deaminase that is a member of the cytidine deaminase family. The protein is involved in somatic hypermutation, gene conversion, and class-switch recombination of immunoglobulin genes. Defects in this gene are the cause of autosomal recessive hyper-IgM immunodeficiency syndrome type 2 (HIGM2).
Specifications
| AID | |
| Polyclonal | |
| Unconjugated | |
| Aicda | |
| activation induced deaminase; activation-induced cytidine deaminase; Aicda; AID; Arp2; CDA2; Cytidine aminohydrolase; epididymis secretory protein Li 284; HEL-S-284; HIGM2; integrated into Burkitt's lymphoma cell line Ramos; single-stranded DNA cytosine deaminase | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11628 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.58 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9WVE0 | |
| Aicda | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 17-130 of human AICDA (NP_0657121). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction