missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AID Polyclonal Antibody

Product Code. 12867732
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12867732 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12867732 Supplier Invitrogen™ Supplier No. PA520012

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is Ramos cell lysate. PA5-20012 can be used with blocking peptide PEP-0131.

This gene encodes a RNA-editing deaminase that is a member of the cytidine deaminase family. The protein is involved in somatic hypermutation, gene conversion, and class-switch recombination of immunoglobulin genes. Defects in this gene are the cause of autosomal recessive hyper-IgM immunodeficiency syndrome type 2 (HIGM2).
TRUSTED_SUSTAINABILITY

Specifications

Antigen AID
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Aicda
Gene Accession No. Q9GZX7, Q9WVE0
Gene Alias activation induced deaminase; activation-induced cytidine deaminase; Aicda; AID; Arp2; CDA2; Cytidine aminohydrolase; epididymis secretory protein Li 284; HEL-S-284; HIGM2; integrated into Burkitt's lymphoma cell line Ramos; single-stranded DNA cytosine deaminase
Gene Symbols Aicda
Host Species Rabbit
Immunogen A peptide corresponding to 14 amino acids near the carboxy terminus of human AID.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11628, 57379
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.