Learn More
AID Monoclonal Antibody (mAID-2), Biotin, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 13-5959-82
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Description: The monoclonal mAID-2 recognizes human and mouse AID (gene AICDA) also known as Activation-Induced Cytidine Deaminase. AID is a 24 kDa RNA-editing enzyme that converts cytosine into uracil. It plays a critical role during B cell development, specifically in somatic hypermutation (SHM) and immunoglobulin class switch recombination (CSR). Changes in levels of AID expression typically present with disease. For example, mutations in AID are found in Hyper-IgM Syndrome (type 2), a disease form without opportunistic infections, while overexpression correlates with poor prognosis in chronic lymphocytic leukemia/small lymphocytic lymphoma. AID is expressed mainly by activated mature B cells such as mature germinal centre (GC) B cells but also in intermediate GC cells (defined as IgD+CD38-CD23-CD71+). In addition to hematopoietic cells, AID is expressed in primary human hepatocellular carcinomas, helicobacter infected gastric epithelial cells, oocytes and embryonic stem cells although it role in these cells is not well understood. Because AID contains an NLS (nuclear localization signal), expression can be found both in the nucleus as well as in the cytoplasm. Applications Reported: This mAID-2 antibody has been reported for use in western blotting, immunohistochemical staining of frozen tissue sections, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, and microscopy.
This gene encodes a RNA-editing deaminase that is a member of the cytidine deaminase family. The protein is involved in somatic hypermutation, gene conversion, and class-switch recombination of immunoglobulin genes. Defects in this gene are the cause of autosomal recessive hyper-IgM immunodeficiency syndrome type 2 (HIGM2).Specifications
AID | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q9GZX7, Q9WVE0 | |
Aicda | |
Affinity chromatography | |
RUO | |
11628, 57379 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
mAID-2 | |
Biotin | |
Aicda | |
activation induced deaminase; activation-induced cytidine deaminase; Aicda; AID; Arp2; CDA2; Cytidine aminohydrolase; epididymis secretory protein Li 284; HEL-S-284; HIGM2; integrated into Burkitt's lymphoma cell line Ramos; single-stranded DNA cytosine deaminase | |
Rat | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2a κ |
For Research Use Only.