missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AICDA Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-93454
This item is not returnable.
View return policy
Description
AICDA Polyclonal antibody specifically detects AICDA in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence, ELISA.
Specifications
| AICDA | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| activation-induced cytidine deaminase, AIDCytidine aminohydrolase, ARP2, CDA2, HIGM2EC 3.5.4.5, integrated into Burkitt's lymphoma cell line Ramos | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Predicted cross-reactivity based on sequence identity: Pig and Cow. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.1 - 0.3 ug/ml, Immunocytochemistry/Immunofluorescence, Peptide ELISA Detection limit 1:1000 | |
| P00441 | |
| AICDA | |
| Peptide with sequence RRKFLYQFKNVR-C corresponding to N-Terminus according to NP_065712.1. | |
| 0.1 mg | |
| Chromatin Modifiers, DNA Repair, Epigenetics, Immunology, Lipid and Metabolism | |
| 57379 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction