missing translation for 'onlineSavingsMsg'
Learn More

AICDA Antibody, Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18415913 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18415913 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18415913 Supplier Novus Biologicals Supplier No. NBP177235AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AICDA Polyclonal antibody specifically detects AICDA in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen AICDA
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias activation-induced cytidine deaminase, AIDCytidine aminohydrolase, ARP2, CDA2, HIGM2EC 3.5.4.5, integrated into Burkitt's lymphoma cell line Ramos
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 14 amino acids near the carboxy terminus of human AID. The immunogen is located within the last 50 amino acids of AID. Amino Acid Squence: EVDDLRDAFRTLGL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Repair, Epigenetics, Immunology, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 57379
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.