missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ AICDA 293T Cell Transient Overexpression Lysate (Denatured)
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Specifications
Specifications
| Cell Line | 293T |
| For Use With (Application) | Western Blot |
| Description | AICDA 293T Cell Transient Overexpression Lysate(Denatured) |
| Gene Accession No. | NM_020661.1 |
| Molecular Weight (g/mol) | Theoretical M.W. 24kDa |
| Plasmid | pCMV-AICDA full-length |
| Quality Control Testing | Transient overexpression cell lysate was tested with anti-AICDA antibody (Mfr. No. H00057379-D01P) by Western Blot. |
| Quantity | 100 μL |
| Storage Buffer | 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) |
| Host Species | Human |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction