missing translation for 'onlineSavingsMsg'
Learn More

AICAR, MedChemExpress

Product Code. p-200096595
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16491076

Brand: Medchem Express HY13417

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AICAR (Acadesine) is an adenosine analog and a AMPK activator. AICAR regulates the glucose and lipid metabolism, and inhibits proinflammatory cytokines and iNOS production. AICAR is also an autophagy, YAP and mitophagy inhibitor.

AICAR (Acadesine) is an adenosine analog and a AMPK activator. AICAR regulates the glucose and lipid metabolism, and inhibits proinflammatory cytokines and iNOS production. AICAR is also an autophagy, YAP and mitophagy inhibitor.

Specifications

Chemical Name or Material AICAR
CAS 2627-69-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C9H14N4O5
Quantity 200 mg
Synonym Acadesine AICA Riboside
Solubility Information DMSO : 125 mg/mL (484.06 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 65 mg/mL (251.71 mM; Need ultrasonic and warming)
SMILES OC[C@@H]1[C@@H](O)[C@@H](O)[C@H](N2C=NC(C(N)=O)=C2N)O1
Molecular Weight (g/mol) 258.23
Formula Weight 258.23
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.