missing translation for 'onlineSavingsMsg'
Learn More

AICAR, MedChemExpress

Product Code. 16441837
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16441837 50 mg 50mg
16491076 200 mg 200mg
16486176 100 mg 100mg
16441666 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16441837 Supplier Medchem Express Supplier No. HY13417

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AICAR (Acadesine) is an adenosine analog and a AMPK activator. AICAR regulates the glucose and lipid metabolism, and inhibits proinflammatory cytokines and iNOS production. AICAR is also an autophagy, YAP and mitophagy inhibitor.

AICAR (Acadesine) is an adenosine analog and a AMPK activator. AICAR regulates the glucose and lipid metabolism, and inhibits proinflammatory cytokines and iNOS production. AICAR is also an autophagy, YAP and mitophagy inhibitor.

Specifications

Chemical Name or Material AICAR
CAS 2627-69-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C9H14N4O5
Quantity 50 mg
Synonym Acadesine AICA Riboside
Solubility Information DMSO : 125 mg/mL (484.06 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 65 mg/mL (251.71 mM; Need ultrasonic and warming)
SMILES OC[C@@H]1[C@@H](O)[C@@H](O)[C@H](N2C=NC(C(N)=O)=C2N)O1
Molecular Weight (g/mol) 258.23
Formula Weight 258.23
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.