missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies AICAR

Product Code. 17130112
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17130112 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17130112 Supplier STEMCELL Technologies Supplier No. 72704
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

AMPK activator

  • AICAR is an adenosine analog that selectively activates AMP-activated protein kinase (AMPK)
  • AMPK regulates lipid and glucose metabolism to maintain cellular energy homeostasis (Hardie and Carling)

Specifications

CAS 2627-69-2
Synonym AICA Riboside; Acadesine; NSC 105823
Quantity 50 mg
Molecular Formula C9H14N4O5
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.