missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIBZIP Antibody (PCRP-CREB3L4-1A3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23702-100ug
This item is not returnable.
View return policy
Description
AIBZIP Monoclonal antibody specifically detects AIBZIP in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| AIBZIP | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry : 1-2 μg/million cells, Immunocytochemistry/ Immunofluorescence : 1-2 μg/mL, Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| AIbZIP, AIBZIPcAMP-responsive element-binding protein 4, Androgen-induced basic leucine zipper protein, ATCE1JAL, Attaching to CRE-like 1, cAMP responsive element binding protein 1, cAMP responsive element binding protein 3-like 4, CREB3, CREB4CREB-4, cyclic AMP-responsive element-binding protein 3-like protein 4, Cyclic AMP-responsive element-binding protein 4, hJALcAMP-responsive element-binding protein 3-like protein 4, Tisp40, Transcript induced in spermiogenesis protein 40 | |
| Recombinant fragment (aa1-298) of human AIBZIP protein | |
| 100 μg | |
| Neuroscience | |
| 148327 | |
| Store at 4°C. | |
| IgG2c κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| PCRP-CREB3L4-1A3 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction