missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIBZIP Antibody (PCRP-CREB3L4-1A3) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-24138JF585
This item is not returnable.
View return policy
Description
AIBZIP Monoclonal antibody specifically detects AIBZIP in Human samples. It is validated for Flow Cytometry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| AIBZIP | |
| Monoclonal | |
| Janelia Fluor 585 | |
| AIbZIP, AIBZIPcAMP-responsive element-binding protein 4, Androgen-induced basic leucine zipper protein, ATCE1JAL, Attaching to CRE-like 1, cAMP responsive element binding protein 1, cAMP responsive element binding protein 3-like 4, CREB3, CREB4CREB-4, cyclic AMP-responsive element-binding protein 3-like protein 4, Cyclic AMP-responsive element-binding protein 4, hJALcAMP-responsive element-binding protein 3-like protein 4, Tisp40, Transcript induced in spermiogenesis protein 40 | |
| Recombinant fragment (aa1-298) of human AIBZIP protein | |
| 0.1 mL | |
| Neuroscience | |
| 148327 | |
| Store at 4°C in the dark. | |
| IgG2c κ |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| PCRP-CREB3L4-1A3 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction