missing translation for 'onlineSavingsMsg'
Learn More

AIB-1 Mouse, Unlabeled, Clone: 34, BD

Product Code. 15825799
Change view
Click to view available options
Quantity:
150 μg
50 μg
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15825799 50 μg Each
15835799 150 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15825799 Supplier BD Biosciences Supplier No. 611104

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Signal transduction via nuclear hormone receptors is important for cell growth and differentiation, development, and homeostasis. Nuclear hormone receptors are ligand-activated transcription factors that modulate target gene expression. These ligand/receptor complexes also interact with transcriptional coactivators which enhance ligand-dependent transcription. Various classes of coactivators have been identified, including SRC-1 and its related proteins, such as TIF-2/GRIP-1, RIP140, AIB-1, and TIF-1α and -1β. AIB-1 (Amplified In Breast cancer-1) is also known as pCIP, RAC3, and TRAM-1. It interacts with estrogen receptor (ER) and is overexpressed in breast cancer biopsies and several breast and ovarian cancer cell lines. Similar to SRC-1 and TIF2, AIB-1 contains a basic helix-loop-helix (bHLH) domain followed by a PAS (Per/Arnt/Sim) region, serine and threonine rich regions, and a charged cluster. In addition, AIB-1 contains three copies of the conserved LXXLL motif which is critical to its interaction with the nuclear receptor. Thus, AIB-1 is a coactivator of nuclear receptors that may participate in the development of steroid-dependent cancers.

TRUSTED_SUSTAINABILITY

Specifications

Antigen AIB-1
Applications Western Blot
Classification Monoclonal
Clone 34
Concentration 250μg/mL
Conjugate Unconjugated
Description Amplified In Breast cancer-1; pCIP; RAC3; TRAM-1
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human AIB-1 aa. 376-389
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.