missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ AI/AS 1610 Liquid Autosampler

Product Code. 31903477 Shop All Thermo Scientific Products
Change view
Click to view available options
Sample Capacity:
155 x 2 mL Vials
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sample Capacity unitSize
31903477 155 x 2 mL Vials Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31903477 Supplier Thermo Scientific™ Supplier No. MI2510000003
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

AI/AS 1610 Liquid Autosampler with 155 vial capacity, enhances liquid sampling automation with high throughput, and automated injection parameter optimization for reliable and efficient GC and GC-MS workflows.

  • 155-vial capacity autosampler designed for high-throughput GC and GC-MS workflows
  • Designed for robust, reliable performance in high-throughput and QA/QC laboratory environments
  • Self-aligning system ensures fast setup without manual position adjustment, supporting both new and experienced users
  • Automatic recognition of injector type and injection mode optimizes injection parameters for safe and reliable operation
  • Wide injection volume range from 0.01 to 80 μL supports both low and high volume applications
  • Fast injection capability (less than 100 ms) ensures accurate and reproducible sample introduction
  • Supports standard 2 mL vials and optional micro-volume vials (300 μL)
  • Compatible with screwed, crimped, and snap-capped vials for flexible sample preparation workflows
  • Advanced syringe washing with up to four solvents reduces carryover and improves analytical accuracy
  • Illuminated syringe compartment with magnifying lens enables easy syringe inspection and replacement
  • Integrated connectivity with a single cable for communication, synchronization, and power supply
  • Seamless integration with Thermo Scientific™ Chromatography Data Systems (Chromeleon, TraceFinder, Xcalibur)
  • Dual-Tower Gemini configuration for simultaneous injection on two channels to increase productivity and throughput available on request
TRUSTED_SUSTAINABILITY

Specifications

Type Autosampler
Sample Capacity 155 x 2 mL Vials
Injection Mode 8 Fully Customizable Method-Specific Injection Mode Presets
Injection Speed Fast/Slow/Variable (1 to 60,000.00 μL/min.)
Injection Volume 0.01 to 80 μL
For Use With (Equipment) TRACE 1600 Series
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.