missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AHSP Polyclonal Antibody

Description
Immunogen sequence: ALLKANKDLI SAGLKEFSVL LNQQVFNDPL VSEEDMVTVV EDWMNFYINY YRQQVTGEPQ ERDKALQELR QELNTLANPF LAKYRDFLKS H Highest antigen sequence identity to the following orthologs: Mouse - 28%, Rat - 66%.
Specifications
Specifications
| Antigen | AHSP |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | AHSP |
| Gene Accession No. | Q9NZD4 |
| Gene Alias | AHSP; alpha hemoglobin stabilising protein; alpha hemoglobin stabilizing protein; alpha-hemoglobin-stabilizing protein; Edrf; Eraf; erythroid associated factor; erythroid differentiation associated factor; erythroid differentiation-related factor; Erythroid-associated factor |
| Gene Symbols | AHSP |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?