missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AHSA1 Antibody (SR1727), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22144-50ul
This item is not returnable.
View return policy
Description
AHSA1 Monoclonal antibody specifically detects AHSA1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| AHSA1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunoprecipitation | |
| SR1727 | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:40 | |
| activator of 90 kDa heat shock protein ATPase homolog 1, AHA1, activator of heat shock 90kDa protein ATPase homolog 1 (yeast), C14orf3, chromosome 14 open reading frame 3, p38AHA1 | |
| A synthesized peptide derived from human AHSA1 (Uniprot #: O95433) | |
| 50 μg | |
| Signal Transduction | |
| 10598 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction