missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ AHSA1 Antibody (1A2-A8), Novus Biologicals™

Product Code. 18346648 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18346648 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18346648 Supplier Novus Biologicals™ Supplier No. H00010598M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

AHSA1 Monoclonal antibody specifically detects AHSA1 in Human,Mouse,Rat samples. It is validated for Western Blot,ELISA,Sandwich ELISA,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen AHSA1
Applications Western Blot, ELISA, Sandwich ELISA, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 1A2-A8
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH00321
Gene Alias activator of 90 kDa heat shock protein ATPase homolog 1, AHA1, activator of heat shock 90kDa protein ATPase homolog 1 (yeast), C14orf3, chromosome 14 open reading frame 3, p38AHA1
Host Species Mouse
Immunogen AHSA1 (AAH00321, 1 a.a. ∽ 338 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAKWGEGDPRWIVEERANATNVNNWHWTERDASNWSTDKLKTLFLAVQVQNEEGKCEVTEVSKLDGEASINNRKGKLIFFYEWSVKLNWTGTSKSGVQYKGHVEIPNLSGENSVDEVEISVSLAKDEPDTNLVALMKEEGVKLLREAMGIYISTLKTEFTQGMILPTMNGESVDPVGQPALKTEERKAKPAPSKTQARPVGVKIPTCKITLKETFLTSPEELYRVFTTQELVQAFTHAPATLEADRGGKFHMVDGNVSGEFTDLVPEKHIVMKWRFKSWPEGHFATITLTFIDKNGETELCMEGRGIPAPEEERTRQGWQRYYFEGIKQTFGYGARLF
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10598
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.