missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AHR Recombinant Rabbit Monoclonal Antibody (23GB1635), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | AHR |
|---|---|
| Clone | 23GB1635 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089241
![]() |
Invitrogen™
MA553350 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088544
|
Invitrogen™
MA553351 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AHR Recombinant Monoclonal antibody specifically detects AHR in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| AHR | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P35869 | |
| 196 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB1635 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Ah; Ah receptor; Ahh; Ahr; Ahre; AH-receptor; aromatic hydrocarbon receptor; aryl hydrocarbon receptor; aryl-hydrocarbon receptor; bHLHe76; Class E basic helix-loop-helix protein 76; dioxin receptor; In | |
| A synthesized peptide derived from human AhR (724-780AA). | |
| Primary | |
| -20°C | |
| AHR |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title