Learn More
AHR Antibody (RPT9) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 7 publications
Brand:  Novus Biologicals NB300-515
Product Code. 18720414
  • £448.00 / 100µL
Description
AHR Monoclonal specifically detects AHR in Human, Mouse, Rat samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Gel Super Shift Assays.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
AHR
Monoclonal
1 mg/mL
Western Blot 1:500 - 1:2000, Flow Cytometry 1:50, ELISA 1:100 - 1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10 - 1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Gel Super Shift Assays 1:1 - 1:100, Chromatin Immunoprecipitation (ChIP) 1:100
P30561
AHR
Synthetic peptide corresponding to residues R(12) K R R K P(17) V(22) K P I P A E G I K(31) of mouse AhR.
100 μL
Apoptosis, Cancer, Hypoxia, Innate Immunity, Transcription Factors and Regulators
196
Human, Mouse, Rat
Purified
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry
RPT9
Unconjugated
No buffer with 0.05% Sodium Azide
Ah receptor, AhR, AH-receptor, aryl hydrocarbon receptor, BHLHE76, bHLHe76aromatic hydrocarbon receptor, Class E basic helix-loop-helix protein 76
Mouse
Protein A purified
RUO
Primary
Detects the Aryl Hydrocarbon Receptor (AHR).
Store at -20C. Avoid freeze-thaw cycles.
IgG3 κ