missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
AHR Monoclonal antibody specifically detects AHR in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, ChIP assay (ChIP), KnockDown
Specifications
Specifications
| Antigen | AHR |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunoprecipitation, ChIP Assay, KnockDown |
| Classification | Monoclonal |
| Clone | 3B12 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, ELISA 0.03ng/mL, Immunocytochemistry/ Immunofluorescence 20μg/mL, Immunoprecipitation 1:10 to 1:500, Chromatin Immunoprecipitation (ChIP), Knockdown Validated |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | NP_001612 |
| Gene Alias | Ah receptor, AhR, AH-receptor, aryl hydrocarbon receptor, BHLHE76, bHLHe76aromatic hydrocarbon receptor, Class E basic helix-loop-helix protein 76 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?