missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
AHNAK Monoclonal antibody specifically detects AHNAK in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen)
Specifications
Specifications
| Antigen | AHNAK |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Frozen) |
| Formulation | 50mM Sodium Borate |
| Research Discipline | Neuroscience |
| Target Species | Human, Mouse |
| Form | Purified |
| Isotype | IgG1 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?