missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AHCYL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10658-3-AP-20UL
This item is not returnable.
View return policy
Description
AHCYL1 belongs to the adenosylhomocysteinase family. The protein, an inositol 1, 4, 5-trisphosphate receptor-binding protein, specifically binds to and activates pancreas-type Na+/HCO3- cotransporter 1 (pNBC1). The regulation through AHCYL1 enables NBC1 variants to have different physiological roles.Specifications
| AHCYL1 | |
| Polyclonal | |
| Unconjugated | |
| AHCYL1 | |
| AdoHcyase 2, AHCYL1, DCAL, IRBIT, PRO0233, XPVKONA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10768, 229709, 362013 | |
| -20°C | |
| Liquid |
| Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| B5DFN2, O43865, Q80SW1 | |
| Ahcyl1 | |
| AHCYL1 Fusion Protein Ag1017 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction