missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AHCYL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38463-100ul
This item is not returnable.
View return policy
Description
AHCYL1 Polyclonal antibody specifically detects AHCYL1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| AHCYL1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| adenosylhomocysteinase-like 1, AdoHcyase 2, DCAL, DC-expressed AHCY-like molecule, dendritic cell expressed AHCY-like protein, inositol 14,5-trisphosphate receptor-binding protein, IRBIT, PRO0233, putative adenosylhomocysteinase 2, S-adenosyl homocysteine hydrolase homolog, S-adenosylhomocysteine hydrolase-like 1, S-adenosylhomocysteine hydrolase-like protein 1, S-adenosyl-L-homocysteine hydrolase 2, XPVKONAEC 3.3.1.1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 31-260 of human AHCYL1 (NP_006612.2).,, Sequence:, MATVTKAPKKQIQFADDMQEFTKFPTKTGRRSLSRSISQSSTDSYSSAASYTDSSDDEVSPREKQQTNSKGSSNFCVKNIKQAEFGRREIEIAEQDMSALISLRKRAQGEKPLAGAKIVGCTHITAQTAVLIETLCALGAQCRWSACNIYSTQNEVAAALAEAGVAVFAWKGESEDDFWWCIDRCVNMDGWQANMILDDGGDLTHWVYKKYPNVFKKIRGIVEESVTGVH | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10768 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction